Enzastaurin 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-(1-Methyl-1H-indol-3-yl)-4-(1-(1-(pyridin-2-ylmethyl)piperidin-4-yl)-1H-indol-3-yl)-1H-pyrrole-2,5-dione
J67480-MA
J67480-MB
J67480-MF
CAAAJ67480-MAEA
235.42
CAD
CAAAJ67480-MA
CAAAJ67480-MB
CAAAJ67480-MF
Enzastaurin 99%
Enzastaurin
Enzastaurin, 99%. Powder.
Purity: 99%
CAS No.: 170364-57-5
Molecular Formula: C32H29N5O2
Formula Weight: 515.60
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: Red to orange
Melting Point: 249-261°
Formula:
C₃₂H₂₉N₅O₂ MW: 515.617 g/mol Melting Pt: 249…261 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD11040980 CAS Number: 170364-57-5 Merck Index: 14,10293 |
Specification Test Results
Assay Minimum | 99 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...