(2'Z,3'E)-6-Bromoindirubin-3'-oxime 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
J67275-MA
J67275-MF
CAAAJ67275-MAEA
244.9
CAD
CAAAJ67275-MA
CAAAJ67275-MF
(2'Z,3'E)-6-Bromoindirubin-3'-oxime 97%
(2'Z,3'E)-6-Bromoindirubin-3'-oxime
6-Bromoindirubin-3'-oxime, 97%. Solid.
Purity: 97%
CAS No.: 667463-62-9
Molecular Formula: C16H10BrN3O2
Formula Weight: 356.17
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Sensitivity: Air and Light Sensitive
Appearance: Dark red
MDL No.: MFCD08705318
Formula:
C₁₆H₁₀BrN₃O₂ Storage Temperature: Ambient |
MDL Number:
MFCD08705318 CAS Number: 667463-62-9 |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...