Potassium chloride 99%, powder for molecular biology
Supplier: Thermo Scientific Chemicals
Synonyms:
Potassiumchloride
J64189-36
J64189-A1
CAAAJ64189-36EA
243.32
CAD
CAAAJ64189-36
CAAAJ64189-A1
Potassium chloride 99%, powder for molecular biology
Potassium chloride
Potassium chloride, 99%, Molecular Biology Grade. Powder.
CAS No.: 7447-40-7
Molecular Formula: KCl
Formula Weight: 74.55
Physical Form: Powder
Density: 1.984
Appearance: White
Odor: Odorless
Melting Point: 773°
Boiling Point: 1500° subl.
MDL No.: MFCD00011360
Formula:
KCl Boiling Pt: 1500 °C (subl.) Melting Pt: 773 °C Density: 1.984 Storage Temperature: Ambient |
MDL Number:
MFCD00011360 CAS Number: 7447-40-7 EINECS: 231-211-8 Merck Index: 14,7621 |
Specification Test Results
Assay Minimum | 99 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...