Nuclear Fast Red, enhanced stability

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sodium 1-amino-2,4-dihydroxy-9,10-dihydro-9,10-dioxoanthracene-3-sulfonate, CI 60760, Sodium 1-amino-2,4-dihydroxy-9,10-dihydro-9,10-dioxoanthracene-3-sulphonate, Helio Fast Rubin BBL

J61010-AP J61010-AU
CAAAJ61010-APEA 324.55 CAD
CAAAJ61010-AP CAAAJ61010-AU
Nuclear Fast Red, enhanced stability
Nuclear Fast Red

A histological counterstain. Nuclear Fast Red solution turns cell nuclei red and cytoplasm a pale pink.


Physical Form: Liquid Appearance: Red Odor: Odorless

Formula: C₁₄H₈NNaO₇S
Storage Temperature: Ambient
MDL Number: MFCD00078493
CAS Number: 6409-77-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR