UltraTech Gloves, MCR Safety
Supplier: MCR Safety
9683L 9683M 9683XL 9683XS
80081-674PK 152.96 CAD
80081-674 80081-676 80081-680 80081-682
UltraTech Gloves, MCR Safety
Gloves
ULTRA FIT, ULTRA COMFORT and ULTRA PROTECTION are the distinguishing characteristics of the UltraTech® series.
  • The 9683 features a 15 gauge white nylon shell and gray nitrile coated palm and fingers
  • Have CE score of 4121
UltraTech® provides better fit and greater function with different polymers on nylon.
Ordering information: These gloves are polybagged by the pair.