UltraTech Dyneema® Gloves, MCR Safety
Supplier: MCR Safety
9676L 9676M 9676S 9676XL
89087-954PK 878.18 CAD
89087-954 89087-956 89087-958 89087-960 89087-962
UltraTech Dyneema® Gloves, MCR Safety
One of the most distinguishing DSM Dyneema® characteristics is its cool-to-touch property.
  • 9676 UltraTech Dyneema® glove features a 13 gauge certified DSM Dyneema® blended shell with PU coated palm and fingers
  • 9676 carries a CE score of 4343 and ANSI cut level of 2
DSM Dyneema® fiber's inherent ability to dissipate body heat and pass moisture from inside of the glove to the outside, results in longer wear time which promotes greater hand protection.
Ordering information: These gloves are polybagged by the pair.