Dupont™ Kevlar® Gloves, Regular Weight String Knit, Plain Shell, MCR Safety
Supplier: MCR Safety
9370L 9370M 9370S 9370XL
80081-636PK 404.4 CAD
80081-636 80081-638 80081-640 80081-642
Dupont™ Kevlar® Gloves, Regular Weight String Knit, Plain Shell, MCR Safety
Gloves
Kevlar® string knit gloves are used for general purpose work applications to provide cut and abrasion resistance, using high performance yarns, to further protect the hands when working with sharp objects.
  • The 9370 features a 7 gauge regular weight 100% Dupont™ Kevlar® string knit material
  • It has a plain shell
Various material weights provide longer durability or additional dexterity where needed.