NitriShield™, Premium Nitrile Gloves, Powder-Free, MCR Safety
Supplier: MCR Safety
6015L 6015M 6015S 6015XL
80081-523CS 333.84 CAD
80081-523 80081-525 80081-527 80081-529
NitriShield™, Premium Nitrile Gloves, Powder-Free, MCR Safety
Gloves
Nitrile contain no latex proteins and offer excellent resistance to punctures and tears.
  • Blue powder-free nitrile
  • Component materials comply with all federal regulations for food contact
  • Rolled cuff
  • Textured grip
Nitrile gloves are three times more puncture resistant than rubber and can be used to offer superior resistance to many types of chemicals. This glove is an excellent alternative for individuals who experience allergic reactions to latex. The 6015 is a industrial / food service grade powder free nitrile glove that measures 4 mil in thicknessMore Product Information