Neoprene Gloves, Industrial Grade, MCR Safety
Supplier: MCR Safety
5435L 5435M 5435S 5435XL
80081-498PK 220.68 CAD
80081-498 80081-500 80081-502 80081-504
Neoprene Gloves, Industrial Grade, MCR Safety
Gloves
Our neoprene gloves are thick, waterproof, and an effective replacement for latex and vinyl products.
  • Black flock lined neoprene
  • Component materials comply with all federal regulations for food contact
  • Straight cuff
Neoprene is a synthetic rubber that maintains flexibility over a wide range of temperatures.