Latex Canner Gloves, Industrial Grade, MCR Safety
Supplier: MCR Safety
5250L 5250XL
80081-426PK 59.68 CAD
80081-426 80081-428 80081-432
Latex Canner Gloves, Industrial Grade, MCR Safety
Gloves
Dexterity with Flock Lining. The flock lining in the gloves provide for easy on and off plus moisture absorption.
  • Scalloped cuff
The 5250 is a yellow flock lined latex glove that is 15 mil in thickness and 12" long.