MedTech Gloves™, Disposable, Medical Grade Gloves, Textured, Powder-Free, NFPA Certified, MCR Safety
Supplier: MCR Safety
5049L 5049M 5049S 5049XL
80081-371CS 585.93 CAD
80081-371 80081-373 80081-375 80081-377
MedTech Gloves™, Disposable, Medical Grade Gloves, Textured, Powder-Free, NFPA Certified, MCR Safety
Gloves
Rubber latex is, by its very nature, a natural substance.
  • Textured grip
  • Powder free
The natural rubber latex polymer used in the manufacture of natural rubber gloves is a biodegradable material that is inherently antimicrobial and antibacterial. The 5049 features 11 mil thickness, is medical grade (Class 1 medical device).
Certifications: NFPA 1999 certified.
Caution: These products contain natural rubber latex which may cause allergic reactions in some people.