SensaGuard™ Gloves, Disposable, Industrial Grade, Powderfree, MCR Safety
Supplier: MCR Safety
5014L 5014M 5014XL 5015L 5015M 5015S 5015XL
80081-301CS 80.67 CAD
80081-301 80081-303 80081-307 80081-309 80081-311 80081-313 80081-315
SensaGuard™ Gloves, Disposable, Industrial Grade, Powderfree, MCR Safety
Gloves
These industrial grade disposables are suited for lots of applications and can be employed for basic household cleanup tasks, food processing or light industrial work.
  • Reversible
  • Made of vinyl material
  • Latex-free
These could include washing dishes, using common cleaning chemicals or having to put the hands in contact with hair dyes. They do protect the hands from direct exposure to various things that might create illness or cause skin breakout.
Ordering information: These gloves are packaged as 100 per dispenser.