DM140-SC Redox Electrode, METTLER TOLEDO®
Supplier: Mettler-Toledo
89598 89598
CA46610-822EA 1258.32 CAD
46610-822 CA46610-822
DM140-SC Redox Electrode, METTLER TOLEDO®
Electrodes Redox Electrodes
This Redox pH electrode combined with platinum ring and ceramic frit junction, is ideal for a wide variety of redox titrations.
  • Platinum ring
  • Ceramic frit junction
  • ARGENTHAL™ lead-off and silver ion trap
  • S7 screw head
Electrode features ARGENTHAL™ lead-off and silver ion trap for optimum silver ion-free accuracy at various sample temperatures. S7 screw head connector is compatible with various cables, so electrode may be used with a wide range of meters and detachable cable.
Ordering information: Electrode includes a quality certificate and a serial number for full traceability.