Cellotriose 95%, powder

Supplier: Thermo Scientific Chemicals

J67702-LB0 J67702-MA J67702-MB
CAAAJ67702-LB0EA 257.54 CAD
CAAAJ67702-LB0 CAAAJ67702-MA CAAAJ67702-MB
Cellotriose 95%, powder
Cellotriose

A 3-glucose cellulodextrin derived from cellulose degradation. An energy source for cellulosic bacteria. Can be used to help identify, differentiate and characterize oligosaccharide metabolizing enzymes such as endoglucanases and _x001A_-glucosidases. Also useful in saccharification and ethanol fermentation research._x000D_


CAS No.: 33404-34-1 Molecular Formula: C18H32O16 Formula Weight: 504.44 Storage Temperature: -30°C to -10°C Physical Form: Powder Sensitivity: Hygroscopic Appearance: White to off-white Solubility: Soluble in water at 50mg/ml Beilstein Reference: 100353 MDL No.: MFCD00069842

Formula: C₁₈H₃₂O₁₆
Storage Temperature: Refrigerator
MDL Number: MFCD00069842
CAS Number: 33404-34-1

Order Now

Specification Test Results

Assay Minimum 95


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR