Marimastat 98%, solid
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(2R,3S)-N1-((S)-3,3-dimethyl-1-(methylamino)-1-oxobutan-2-yl)-N4,3-dihydroxy-2-isobutylsuccinamide, BB-2516, (2S,3R)-N⁴-((1S)-2,2-Dimethyl-1-((methylamino)carbonyl)propyl)-N¹,2-hydroxy-3-(2-methylpropyl)butanediamide, Marimastat
J67288-MA
J67288-MCR
CAAAJ67288-MAEA
466.1
CAD
CAAAJ67288-MA
CAAAJ67288-MCR
Marimastat 98%, solid
Marimastat
Marimastat, 98%
Purity: 98%
CAS No.: 154039-60-8
Molecular Formula: C15H29N3O5
Formula Weight: 331.41
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White
Melting Point: 148°
MDL No.: MFCD00866242
Formula:
C₁₅H₂₉N₃O₅ MW: 331.413 g/mol Melting Pt: 148 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00866242 CAS Number: 154039-60-8 |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...