IPA-3, solid

Supplier: Thermo Scientific Chemicals
Danger

J65848-LB0 J65848-MA
CAAAJ65848-LB0EA 191.18 CAD
CAAAJ65848-LB0 CAAAJ65848-MA
IPA-3, solid
IPA-3

Acts as an allosteric inhibitor of Pak1


  • Light Sensitive


Yellow solid


Soluble in DMSO or ethanol. Insoluble in water.

Formula: C₂₀H₁₄O₂S₂
Storage Temperature: Refrigerator
MDL Number: MFCD00388917
CAS Number: 42521-82-4
UN: 3077
ADR: "9",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR