Portable ESD Field Service Kit, STATICO
Supplier: Statico
S1920
14223-740EA 330.88 CAD
14223-740
Portable ESD Field Service Kit, STATICO
Static Control Monitors
STATICO's Portable Field Service Kits are designed to protect ESD (Electro Static Discharge) sensitive components and devises at remote locations, where static free workstations are not available
The S1920 is the ideal means for field service operators, safeguarding against ESD damage.
Certifications: Meets ANSI/ESD 20.20-2014.