Double Stack Connector, Statico
Supplier: Statico
S1092-38MFB
14223-736EA 36.86 CAD
14223-736
Double Stack Connector, Statico
Grounders Grounding Wires
Molded male/female double-stack snap with banana jack and caps. Provides a connecting point for:
  • Wrist grounding strap via banana jack
  • Snaps-on table mat either via female or male 3/8" (10 mm) socket
Either one of the unused snaps can be covered by a plastic cap, to prevent shock due to exposure.