Septihol® WFI Sterile Alcohol Solutions
Supplier: Steris
6250E7 625077WR 625096WR
CA89131-736CS 1085.36 CAD
CA89131-736 CA75780-020 CA75780-022
Septihol® WFI Sterile Alcohol Solutions
Cleaning Agents Detergents and Surface Cleansers
STERIS' Septihol® WFI Sterile Alcohol Solution removes residue from hard surfaces even from the most critical surfaces. It can be used straight out of the aerosol can and be sprayed in any direction. Since it contains no stabilizers or denaturants, it evaporates quickly and completely.
  • Productivity - Ready to use sterile alcohol eliminates in-house production validation and documentation concerns
  • Flexibility - Bag on valve aerosol design eliminates contact between product and pressurizing nitrogen, permits operation at any angle, and allows maximum product discharge which minimizes contamination potential and waste while maximizing use flexibility
  • Ease of Use - Double bagged for ease of introduction into aseptic areas
  • Safety - 0.2 µm filtered, clean room grade B (Class 100) filled and packaged minimizes potential for particles being introduces during use
  • Reliability - Each lot is tested to ensure sterility and low endotoxins
STERIS' Septihol® WFI Sterile alcohol solution is an innovative bag on valve aerosol can containing a mixture of 70% (v/v) USP isopropanol 30% USP Water For Injection (WFI). All cans are filled and double bagged in a Grade B (Class 100) facility and gamma irradiated to ensure sterility for easy introduction in critical areas. Each lot is documented by Certificates of Analysis that contain endotoxin, irradiation and sterility results.