One Solution st Water For Injections
Supplier: Steris
One Solution
1S0308WR 1S03E4WR 1S03E4WR 1S0308WR
CA75780-026CS 275.01 CAD
10755-278 CA75780-026 CA75780-024 10755-280
One Solution st Water For Injections
Cleaning Agents Detergents and Surface Cleansers
A diluent for surface sterilants, disinfectants, sanitizers, or cleaners used in cleanrooms or other critical environments; meets USP specifications and is double bagged for easy entry into cleanrooms or aseptic areas.
  • Quality - Extremely low level of endotoxins from WFI produced by a Finn-Aqua® Multiple Effect Water Still
  • Convenience - Gamma irradiated and double bagged for easy introduction into your aseptic areas
  • Assurance - Each lot documented by a Certificate of Analysis including irradiation and sterility resultstested
STERIS' One Solution st WFI is intended to dilute concentrated cleaners or disinfectants.
The family of One Solution st Water For Injection products meets United States Pharmacopeia (USP) Water For Injection (WFI) specifications and is intended to be used to dilute surface disinfectants and cleaners. Sterile filtered and filled in a controlled environment, each package is double bagged and gamma irradiated for easy entry into your controlled environments and aseptic core areas. These products have an extensive documentation package to meet your validation objectives and are supported by the STERIS Technical Services team for individualized application and validation assistance.