Jurkat Cell Lysate, Enzo Life Sciences
Supplier: Enzo Life Sciences
LYC-JK100-F
89141-952EA 226 CAD
89141-952
Jurkat Cell Lysate, Enzo Life Sciences
Cells
Liquid
In SDS-PAGE sample buffer, pH 6.8, heated for 10 min. at 70°C and containing 40mM TRIS-HCl, 1% SDS, 50mM DTT, 7.5% glycerol, and 0.003% bromophenol blue.