Hypochlorite WFI Sterile Solutions
Supplier: Steris
1S0677WR 1S0877WR 1S0896WR 1S0808WR 1S0696WR 1S0608WR
CA75780-028CS 857.39 CAD
CA75780-028 CA75780-036 CA75780-038 CA75780-040 CA75780-056 CA75780-058
Hypochlorite WFI Sterile Solutions
Cleaning Agents Detergents and Surface Cleansers
STERIS' 0.525% or 5.25% Hypochlorite WFI sterile solution removes residue from hard surfaces even from the most critical surfaces. Oxidizing cleaners can be extremely effective with tough soils.
  • Quality - Extremely low level of endotoxins from WFI produced by a Finn-Aqua® Multiple Effect Water Still
  • Effective - High quality sodium hypochlorite solution oxidizes difficult to remove soils
  • Convenience - Double bagged for easy introduction into your aseptic areas
  • Safety - 0.2 micron filtered, aseptically filled and packaged minimizes potential for particles being introduced during use
  • Assurance - Each lot documented by a Certificate of Analysis including sterility results
STERIS' 0.525% or 5.25% Hypochlorite WFI sterile solution is made with United States Pharmacopeia (USP) quality Water For Injection and Sodium Hypochlorite. It is designed to remove residues from surfaces. Sterile filtered and filled in an ISO Class 5 aseptic environment, each package is double bagged for easy entry into your controlled environments. This product has an extensive documentation package to meet your validation objectives and is supported by the STERIS Technical Services team for individualized application and validation assistance.