Veterinary/Laboratory Blood Collection Tubes, Air-Tite
Supplier: Air-Tite
MV3P MV6P MV3R
10147-204CS 308.77 CAD
10147-204 10147-202 10147-206
Veterinary/Laboratory Blood Collection Tubes, Air-Tite
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
Air-Tite Products blood collection tubes are for use in veterinary and laboratory environments.
  • Red top
  • No additives
  • Various size options available
These tubes provide a great value at an affordable cost. Both glass and plastic options are available.
Packaging: Sterile packaging.
Caution: For aboratory and Veterinary Use Only