Anti-H3F3A Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-P10 600-401-P10
CA600-401-P10EA 871.01 CAD
RL600-401-P10 CA600-401-P10
Anti-H3F3A Rabbit Polyclonal Antibody
Antibodies
Anti-Histone H3 K79-Me1 antibody is has been tested for use in Western Blot, ChIP, Immunofluorescence, and Dot Blot. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately ~15kDa.

Recommended Dilutions: ChIP: 2 µg; Immunofluorescence: 1:50 - 1:100; Western Blot: 0.5µg/ml

Type: Primary
Antigen: Histone H3 (HIST2H3C)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: C. elegans, Drosophila, Human, Mouse, Plant, Primate, Rat, Xenopus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR