Anti-AKT2 Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 11F6.B2]

Supplier: Rockland Immunochemicals
200-508-J35 200-508-J35
CA200-508-J35EA 871.01 CAD
RL200-508-J35 CA200-508-J35
Anti-AKT2 Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 11F6.B2]
Antibodies
Anti-AKT2 PE Antibody is suitable for Flow Cytometry, ELISA, immunochemistry, and western blotting. Expect a band approximately 56 kDa in size corresponding to AKT2 Protein by western blotting in the appropriate cell lysate or extract.

Recommended Dilutions:
Restore with deionized water (or equivalent); 100 µl to 20 µl
Do not freeze! Light sensitive!

Type: Primary
Antigen: AKT2
Clonality: Monoclonal
Clone: 11F6.B2
Conjugation: RPE (R-Phycoerythrin)
Epitope: Center
Host: Rat
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR