Anti-PTPRC Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: UCHL1]

Supplier: Rockland Immunochemicals
200-326-N71 200-326-N71
CA200-326-N71EA 871.01 CAD
RL200-326-N71 CA200-326-N71
Anti-PTPRC Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: UCHL1]
Antibodies
Anti-CD45RO is useful for Flow Cytometry, Immunoprecipitation, immunochemistry, Immunofluorescence, and Western Blot. Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 5 µl/test/1x10e5 to 1x10e8 cells;
Do not freeze!

Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: UCHL1
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR