Anti-PTPRC Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 4F2.D9.C5.G5.B2.C5]

Supplier: Rockland Immunochemicals
200-302-N86 200-302-N86
CA200-302-N86EA 754.01 CAD
RL200-302-N86 CA200-302-N86
Anti-PTPRC Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 4F2.D9.C5.G5.B2.C5]
Antibodies
Anti-CD45.2 is useful for immunochemistry, Immunoprecipitation, Immunofluorescence, and Flow Cytometry using mouse spleen cells, or an appropriate cell type (where indicated).

Recommended Dilutions: Flow Cytometry: 10 µl/10^6 cells (0.1 µg); Sufficient to run approximately 500 tests
Do not freeze!

Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: 4F2.D9.C5.G5.B2.C5
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR