Anti-CD4 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 4C8.H6.D4]

Supplier: Rockland Immunochemicals
200-302-M63 200-302-M63
CA10036-874EA 381 CAD
RL200-302-M63 CA10036-874
Anti-CD4 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 4C8.H6.D4]
Antibodies
Anti-CD4 is useful for Immunofluorescence and Flow Cytometry (Cell Surface). Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 10 µl/10^6 cells (0.1 µg)
Do not freeze!

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: 4C8.H6.D4
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR