Anti-IgG1 kappa Rat Monoclonal Antibody [clone: RG1K]

Supplier: Rockland Immunochemicals
012-001-330 012-001-330
CA012-001-330EA 406 CAD
CA012-001-330 RL012-001-330
Anti-IgG1 kappa Rat Monoclonal Antibody [clone: RG1K]
Antibodies
Rat IgG1 Kappa isotype control can be utilized as a control or standard reagent in Western Blotting, Flow Cytometry, and ELISA experiments where determination of sample isotype is important. Specific conditions should be optimized by user.

Recommended Dilutions: Flow Cytometry: 1:1000-1:5000;

Type: Control
Antigen: IgG1 kappa
Clonality: Monoclonal
Clone: RG1K
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR