Samco™ Round Base Test Tubes, 12×75 mm, Thermo Scientific
Supplier: Thermo Scientific
12-010 12-005S 12-012 12-011 12-007S 12-014
89498-436CS 3364.31 CAD
89498-436 89498-428 89498-422 89498-432 89498-430 89498-418
Samco™ Round Base Test Tubes, 12×75 mm, Thermo Scientific
Tubes Test Tubes
Samco™ round based 12×75 mm tubes are suitable for most laboratory applications.
  • Available with or without cap
Available in clear polypropylene for chemical resistance and strength, or polystyrene for superior clarity.
Ordering information: Ambient storage and shipping recommended for tubes.
Packaging: Choice of packaging formats: sterile (bulk or individually wrapped) or non sterile (bulk or blister tray).