Samco™ Two-Position Caps, Thermo Scientific
Supplier: Samco
5A 0002 5A 0007 5A 0008 5A 0012 5A 0016
89498-514CS 4809.38 CAD
89498-514 89498-516 89498-518 89498-520 89498-522 89498-524
Samco™ Two-Position Caps, Thermo Scientific
Closures Test/Sample Tube Closures
A range of Two-Position Caps (12mm, 16mm, and 17mm) designed to fit a wide range of Samco™ round based tubes.
Ordering information: Ambient storage and shipping recommended for caps.
Order Now
SPECIFICATIONS
Environmentally Preferable