Metal Half-Cells pH Electrode, METTLER TOLEDO®
Supplier: Mettler Toledo
59904377 59904391 59904408
CA11238-664EA 629.89 CAD
CA11238-664 CA11238-668 CA11238-670
Metal Half-Cells pH Electrode, METTLER TOLEDO®
Electrodes pH Electrodes
Electrode is for routine applications.
  • S7 screw head
  • Compatible with various cables
Platinum electrodes cover the majority of applications, whereas gold electrode is best for highly oxidizing samples. Silver electrodes are primarily used for silver based electrochemistry such as chloride determination.
S7 screw head connector is compatible with various cables, so electrode may be used with a wide range of meters.