Chlorine Test In Freshwater And Seawater, MQuant®, MilliporeSigma
Supplier: MilliporeSigma
1.14670.0001
CA1.14670.0001EA 58.68 CAD
CA1.14670.0001
Chlorine Test In Freshwater And Seawater, MQuant®, MilliporeSigma
Environmental Test Kits
The test can be used for the determination of chlorine in freshwater and seawater
The chlorine concentration can be measured semiquantitatively by visual comparison of the colour of the measurement solution with the colour fields of the colour card.