Lok-Tight™ Contact Plates for Surface Sampling, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
P580 P520R P520 P595
CA76200-278PK 120.54 CAD
CA76200-278 CA76200-282 CA76200-280 CA76200-284
Lok-Tight™ Contact Plates for Surface Sampling, Hardy Diagnostics
Media for Microbiology Prepared Media for Microbiology
Contact Plates with Friction Lids to ensure sterility in your environmental testing program.
  • Tear notch on outer bag for easy opening; no scissors required
  • New moisture control packaging format prevents moisture buildup and ensures safe and sterile transfer
  • Friction lid design keeps lids in place
  • Lok-Tight™ optional locking feature means lids stay secured during sampling and transport
  • Each plate has a grid molded into the bottom of the plate to aid in the counting of microorganisms
  • Certificate of analysis available online for each lot
These contact plates are recommended for use in the cultivation of microorganisms from environmental surfaces.