ReadyRack™ Tryptic Soy Broth (TSB), Disposable Tube Rack
Supplier: HARDY DIAGNOSTICS CA
K82BX K380BX
CA10436-342PK 1893.85 CAD
CA10436-342 CA75873-760
ReadyRack™ Tryptic Soy Broth (TSB), Disposable Tube Rack
Media for Microbiology Prepared Media for Microbiology
Tryptic Soy Broth (TSB) in a ReadyRack™ is for sterility testing and cultivation of microorganisms. TSB is also known as Soybean-Casein Digest Broth, USP.
  • For sterility testing in cleanrooms
  • Can be rinsed with alcohol
  • Avoid contamination
The ReadyRack is a plastic rack designed for cleanroom use and can be rinsed with alcohol.
Simply remove ReadyRack from box outside of cleanroom. Spray down the plastic bag with isopropyl alcohol, and allow to dry. Take into cleanroom and remove plastic bag. Spray down entire ReadyRack with isopropyl alcohol, and allow to dry.
It is a convenient way to bring media into a controlled area while avoiding outside contamination.