MColortest™ Chromate Test, MilliporeSigma
Supplier: MilliporeSigma
1.14402.0001 1.14441.0001
CA1.14402.0001EA 253 CAD
CA1.14402.0001 CA1.14441.0001
MColortest™ Chromate Test, MilliporeSigma
Environmental Test Kits
The test measures chromium(VI) present in the sample as chromate or dichromate ions in wastewater and percolating water, groundwater and surface water, industrial water and drinking water.