MColortest™ Iron Test, MilliporeSigma
Supplier: MilliporeSigma
1.14438.0001 1.14759.0001 1.18458.0002
CA1.14438.0001EA 245 CAD
CA1.14438.0001 CA1.14759.0001 CA1.18458.0002
MColortest™ Iron Test, MilliporeSigma
Environmental Test Kits
The test measures bivalent and trivalent iron in its dissolved form as well as fresh colloidal iron(III) hydroxide in groundwater and surface water, drinking water, industrial water and wastewater.
When the reagents of the MColortest™ test are used up, the kit can be replenished by ordering the appropriate refill pack. Refill packs are available for a range of MColortest™ test kits, making it a particularly economical system.