Samco™ UrineGUARD™ Container Labels, Thermo Scientific
Supplier: Samco
30 3000 30 3000L 30 3500
89498-500CS 722.85 CAD
89498-500 89498-502 89498-504
Samco™ UrineGUARD™ Container Labels, Thermo Scientific
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
Alleviate mess and exposure when handling urine collection and sampling with the Samco™ 24-Hour Urine Container range
  • Range of sizes: 3000mL and 3500mL
  • Amber tinted to protect from light exposure
  • Easy to read, accurate graduations
  • Custom cap and mouth design helps prevent leakage
  • Large carrying handle for ease of transportation
Containers are amber tinted to protect from light exposure, and they are also inert to normal preservatives.
Certifications: FDA Class 1 Exempt Medical Device.
Ordering information: Ambient storage and shipping recommended for containers.