Samco™ Round Base Test Tubes, 17x100mm, Thermo Scientific
Supplier: Samco
17-009S 17-010
89498-468CS 1397.21 CAD
89498-468 89498-472 89498-476 89498-478 89498-484 89498-486
Samco™ Round Base Test Tubes, 17x100mm, Thermo Scientific
Tubes Culture Tubes
Samco™ round based 17×100 mm tubes are suitable for most laboratory applications.
  • Choice of packaging formats: sterile or nonsterile
  • Available with or without cap
  • Polypropylene or polystyrene construction
Available in clear polypropylene for chemical resistance and strength, or polystyrene for superior clarity.
Ordering information: Ambient storage and shipping recommended for tubes.