SP Bel-Art Polypropylene Freezer Boxes, Bel-Art Products, a part of SP
Supplier: Bel-Art
F18852-0017 F18852-0018
CA10753-246CS 801.52 CAD
CA10753-246 CA10753-248
SP Bel-Art Polypropylene Freezer Boxes, Bel-Art Products, a part of SP
Boxes Cryoboxes
Plastic freezer storage boxes are available in two sizes and hold either 1.5 to 2 mL micro tubes / cryo vials (81-place box) or 5 mL conical centrifuge tubes, including Eppendorf® brand (25-place box).
  • Polypropylene construction
  • Durable
  • Clear translucent lids permit easy visibility
The 81-place micro tube / cryo vial box fits standard 2” ultra-low temperature (ULT) freezer racks and the 25-place 5 mL conical centrifuge tube box fits standard 3" ULT freezer racks. Made of durable polypropylene, they can withstand temperatures to –86°C (-187°F). ClearMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable