Wire Feeders, Stainless Steel, Bio-Serv
Supplier: Avantor
K3551
CA89412-162EA 75.27 CAD
CA89412-162 CA89412-164
Wire Feeders, Stainless Steel, Bio-Serv
Animal Supplies Animal Environmental Enrichment
Bio-Serv's Stainless Steel Wire Feeders offer a simple, safe and convenient way to provide a foraging opportunity for a variety of laboratory species
The simplistic design holds an array of food items, such as long stem hay, fresh fruit and vegetables. Made of heavy gauge stainless steel. Easy to hang, fill and clean. Cagewasher safe and autoclavable.
Fill the wire feeders with edible enrichment products such as hay, lettuce, or other fruits and vegetables. To provide novelty, rotate food items and be creative. The Stainless Steel Wire Feeders come with a quick link so you can easily hang them in the cage.