InnoDome™ and InnoWheel™ Mouse Enrichment Devices, Bio-Serv
Supplier: Bio-Serv
S3174 S3175
CA89412-136EA 1411.6 CAD
CA89412-136 CA89412-138
InnoDome™ and InnoWheel™ Mouse Enrichment Devices, Bio-Serv
Animal Supplies Animal Environmental Enrichment
Made of 100% PETE disposable recyclable plastic
The very low profile design fits in most mouse cage systems. Height of dome and wheel together is 3.027". Transparent color provides ease of visibility. A great addition to breeding programs.
Ordering information: These products are NOT designed to be put through the cagewasher or autoclave.