Best Balls Enrichment Devices, Bio-Serv
Supplier: Bio-Serv
K3222 K3219 K3218 K3464 K3468
CA89067-626EA 54.7 CAD
CA89067-626 CA89067-628 CA89067-630 CA89412-132 CA89412-134
Best Balls Enrichment Devices, Bio-Serv
Animal Supplies Animal Environmental Enrichment
Made of hard polyethylene
Extremely sturdy. 10" ball has recessed screw; add water or sand to change rolling behavior. Cagewasher safe.