Rest Stops™ Rodent Environmental Enrichment, Certified, Bio-Serv
Supplier: Bio-Serv
K3393 K3392 K3391
89412-108EA 16.46 CAD
89412-108 CA89412-106 CA89412-104
Rest Stops™ Rodent Environmental Enrichment, Certified, Bio-Serv
Animal Supplies Animal Environmental Enrichment
Rest Stops offer paw relief from wire bottom floors and provide a comfortable resting area
Made of transparent high-temp polycarbonate. Useful to place special needs diets or enrichment treats at cage level. Autoclavable. Cagewasher safe.
Certifications: Certified (Contaminant Screened).