Pup Tents™ for Mice, Certified, Bio-Serv
Supplier: Bio-Serv
K3390 K3600 K3389
CA89412-080EA 14.9 CAD
CA89412-080 CA89412-082 CA89412-084
Pup Tents™ for Mice, Certified, Bio-Serv
Animal Supplies Animal Environmental Enrichment
A shelter for pups and weanlings made of high-temp polycarbonate
Small, low profile design fits in most mouse cagings. Deflects air flow to prevent thermal stress. Autoclavable. Cagewasher safe.
Certifications: Certified (Contaminant Screened).
Order Now
SPECIFICATIONS
Environmentally Preferable