Rodent Liquid Diet, Lieber-DeCarli '82, Shake and Pour, Bio-Serv
Supplier: Bio-Serv
F1259SP F1258SP F1697SP F1259SP
CA89067-600EA 190.64 CAD
CA89067-600 CA89067-602 CA89067-606 89067-600
Rodent Liquid Diet, Lieber-DeCarli '82, Shake and Pour, Bio-Serv
Animal Supplies Animal Feed
Liquid diets are a versatile diet medium that allows the addition of alcohol, medications or test compounds. A convenient way to measure food intake or study nutritional manipulation. Ideal for post-operative recovery.
  • Easy to prepare; no blender needed
  • Add water, shake and pour: Use with Bio-Serv's 160 oz. Shake and Pour Bottle or your own mixing vessel
  • Custom formulations available
  • Nutritionally assayed
  • Powder
  • Shelf Life: Dry Mix, 6 months refrigerated; Prepared, 3 days refrigerated
Ingredients: Corn Syrup Solids, Casein, Olive Oil, Maltodextrin, Cellulose, Mineral Mix, Corn Oil, Suspending Aids, Safflower Oil, Vitamin Mix, L-Cystine, Choline Bitartrate, DL-Methionine.
Ethanol: 355 kcal/L
Nutritional carbohydrate content: 490 kcal/L for Control, Shake and Pour, 135 kcal/L for Ethanol, Shake and Pour, 135 kcal/L for Ethanol and Maltose Dextrin, Shake and Pour
Nutritional fat content: 359 kcal/L
Nutritional protein content: 151 kcal/L
Total caloric profile: 1000 kcal/L