Rodent Liquid Diet, AIN-76, Shake and Pour, Bio-Serv
Supplier: Bio-Serv
F1268SP F1436SP
CA89067-592EA 185.1 CAD
CA89067-592 CA89067-594
Rodent Liquid Diet, AIN-76, Shake and Pour, Bio-Serv
Animal Supplies Animal Feed
Liquid diets are a versatile diet medium that allows the addition of alcohol, medications or test compounds
Ingredients: Corn Syrup Solids, Casein, Corn Oil, Cellulose, Maltodextrin, Mineral Mix, Suspending Aids, Vitamin Mix, DL-Methionine, Choline Bitartrate.
A convenient way to measure food intake or study nutritional manipulation. Ideal for post-operative recovery. Easy to prepare; no blender needed. Add water, shake and pour. Use with Bio-Serv's 160 oz. Shake and Pour Bottle or your own mixing vessel. Custom formulations available. Nutritionally assayed. Powder. Shelf Life: Dry Mix,More Product Information