Rodent Liquid Diet, AIN-76, Bio-Serv
Supplier: Bio-Serv
F1268 F1436
CA89067-588EA 185.1 CAD
CA89067-588 CA89067-590
Rodent Liquid Diet, AIN-76, Bio-Serv
Animal Supplies Animal Feed
Liquid diets are a versatile diet medium that allows the addition of alcohol, medications or test compounds. A convenient way to measure food intake or study nutritional manipulation. Ideal for post-operative recovery.
  • Custom formulations available
  • Nutritionally assayed
  • Powder
  • Shelf Life: Dry Mix, 6 months refrigerated; Prepared, 3 days refrigerated
Ingredients: Corn Syrup Solids, Casein, Maltodextrin, Corn Oil, Cellulose, Mineral Mix, Suspending Aid, Vitamin Mix, DL-Methionine, Choline Bitartrate.
Ethanol: 355 kcal/L
Nutritional carbohydrateMore Product Information