Dustless Precision Pellets® Rodent Purified Diet, Bio-Serv

Supplier: Bio-Serv
F05684 F0071 F0021 F0078 F05301 F0299 F05684 F0021 F0078 F0299 F0075
CA89067-560EA 658.97 CAD
CA89067-560 CA89067-562 CA89067-564 CA89067-566 CA89067-574 CA89067-576 CA89067-578 89067-560 89067-564 89067-574 89067-578 CA89408-190
Dustless Precision Pellets® Rodent Purified Diet, Bio-Serv
Animal Supplies Animal Feed
Precise size and weight pellets generally used in nutritional and behavioral studies.

  • Dustless and fracture free
  • Superior performance in automatic feeders
  • Nutritionally complete purified formula
  • Wide range of weights: 14 mg to 1 g
  • Available in grain-based formula
  • Every batch tested for hardness, weight, and height
  • Custom flavors and colors available upon request
  • Nutritionally assayed
  • Shelf Life: 12 months, cool dry storage

Ingredients: Sucrose, Dextrose, Casein, Corn Oil, Mineral Mix, Cellulose, Corn Syrup, Calcium Silicate, Vitamin Mix, Magnesium Stearate, Choline Bitartrate, DL-Methionine, L-Cystine, Ascorbic Acid, Vitamin E Acetate, tBHQ, (Chocolate Flavoring in F05301 and F0299).

Caloric value: 3.60 kcal/g
Nutritional ash content: 6.5%
Nutritional carbohydrate content: 59.1%
Nutritional fat content: 5.6%
Nutritional fibre content: 4.7%
Nutritional moisture content: <10%
Nutritional protein content: 18.7%
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR