Dustless Precision Pellets® Rodent Grain-Based Diet, Bio-Serv
Supplier: Bio-Serv
F0163 F0165 F0171 F0173 F0165
CA89067-546EA 602.6 CAD
CA89067-546 CA89067-548 CA89067-554 CA89067-558 89067-548
Dustless Precision Pellets® Rodent Grain-Based Diet, Bio-Serv
Animal Supplies Animal Feed
Precise size and weight pellets generally used in nutritional and behavioral studies.
  • Dustless and fracture free
  • Superior performance in automatic feeders
  • Nutritionally complete, banana-flavored, grain-based formula
  • Available in purified formula
  • Wide range of weights: 20 mg to 1 g
  • Every batch tested for hardness, weight and height
  • Custom flavors and colors available upon request
  • Nutritionally assayed
  • Shelf Life: 12 months, cool dry storage
Ingredients: Ground Corn, Banana Flakes, Dehulled Soybean Meal, Corn Gluten Meal, Ground Wheat, Corn Gluten Feed,More Product Information